Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Canx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110499.1 | NP_001103969.1 | 2 | 315 | ||
NM_001110500.1 | NP_001103970.1 | 2 | 283 | |||
NM_007597.3 | NP_031623.1 | 2 | 319 | |||
GenBank mRNA | AK028279.1 | - | 2 | 246 | ||
AK084175.1 | - | 2 | 259 | |||
AK159298.1 | - | 2 | 303 | |||
AK165508.1 | - | 2 | 218 | |||
AK166649.1 | - | 2 | 296 | |||
BC012408.1 | - | 2 | 237 | 3497769 | ||
BC040244.1 | - | 2 | 246 | 2582119 | ||
L18888.1 | - | 1 | 131 | |||
L23865.1 | - | 1 | 71 |