Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076789.1 | NP_001070257.1 | 2, 3 | 265 | ||
NM_001110216.1 | NP_001103686.1 | 3, 4 | 366 | |||
NM_007626.3 | NP_031652.1 | 2, 3 | 266 | |||
GenBank mRNA | AK008792.1 | - | 2, 3 | 301 | ||
AK018349.1 | - | 2, 3 | 212 | |||
AK030366.1 | - | 2, 3 | 266 | |||
AK030442.1 | - | 3, 4 | 319 | |||
AK032975.1 | - | 2, 3 | 222 | |||
BC004707.1 | - | 2, 3 | 265 | 3499489 | ||
X99641.1 | - | 2, 3 | 198 |