Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081635.1 | NP_001075104.1 | 3 | 440 | ||
NM_001081636.1 | NP_001075105.1 | 3 | 485 | |||
NM_007632.2 | NP_031658.1 | 1 | 346 | |||
GenBank mRNA | AK083384.1 | - | 3 | 425 | ||
AK159503.1 | - | 1 | 272 | |||
AK161758.1 | - | 3 | 364 | |||
AK165191.1 | - | 1 | 379 | |||
AK168290.1 | - | 1 | 274 | |||
BC004076.1 | - | 1 | 238 | 3591985 | ||
BC005605.1 | - | 1 | 237 | 3483584 | ||
M86183.1 | - | 1 | 241 |