Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007633.2 | NP_031659.2 | 3 | 289 | ||
GenBank mRNA | AK089950.1 | - | 3 | 221 | ||
AK163046.1 | - | 3 | 278 | |||
AK165658.1 | - | 3 | 279 | |||
BC062152.1 | - | 3 | 316 | 6510163 | ||
BC084588.1 | - | 3 | 333 | 6506897 | ||
BC106191.1 | - | 3 | 289 | 6336006 | ||
BC138660.1 | - | 2 | 122 | 8861680 | ||
BC138662.1 | - | 2 | 122 | 8861683 | ||
X75888.1 | - | 2 | 129 |