Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007659.3 | NP_031685.2 | 4 | 335 | ||
GenBank mRNA | AK030231.1 | - | 4 | 336 | ||
AK051859.1 | - | 4 | 362 | |||
AK135516.1 | - | 4 | 339 | |||
AK168054.1 | - | 4 | 336 | |||
BC005614.1 | - | 3 | 228 | |||
BC024396.1 | - | 4 | 313 | 4977366 | ||
M38724.1 | - | 3 | 256 | |||
U58633.1 | - | 4 | 339 | |||
X16461.1 | - | 3 | 256 |