Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007674.3 | NP_031700.1 | 2 | 676 | ||
GenBank mRNA | AK141681.1 | - | 2 | 90 | ||
BC119158.1 | - | 2 | 664 | 8733907 | ||
L08061.1 | - | 2 | 679 |