Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007677.2 | NP_031703.1 | 2 | 533 | ||
GenBank mRNA | BC100345.1 | - | 2 | 530 | 30715425 | |
CN703708.1 | - | 2 | 533 | |||
M83344.1 | - | 2 | 549 |