Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007694.4 | NP_031720.1 | 2 | 541 | ||
GenBank mRNA | AK019893.1 | - | 2 | 151 | ||
AK077487.1 | - | 2 | 107 | |||
AK077489.1 | - | 2 | 149 | |||
AK133629.1 | - | 2 | 149 | |||
AK133739.1 | - | 2 | 149 | |||
AK133786.1 | - | 2 | 151 | |||
AK158857.1 | - | 2 | 542 | |||
BC014736.1 | - | 2 | 142 | 4511248 | ||
X51429.1 | - | 2 | 137 | |||
X53028.1 | - | 2 | 147 |