Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042534.3 | NP_001035999.1 | 5 | 663 | ||
NM_001271395.1 | NP_001258324.1 | 5 | 656 | |||
NM_001271415.1 | NP_001258344.1 | 6 | 653 | |||
NM_007599.4 | NP_031625.2 | 5 | 510 | |||
GenBank mRNA | AK146622.1 | - | 5 | 514 | ||
AK149857.1 | - | 5 | 460 | |||
AK152818.1 | - | 5 | 488 | |||
AK159577.1 | - | 5 | 463 | |||
AK159736.1 | - | 5 | 465 | |||
AK165153.1 | - | 5 | 477 | |||
AK165586.1 | - | 3 | 306 | |||
BC003480.1 | - | 5 | 464 | 2811050 | ||
BC023101.1 | - | 5 | 452 | 3983574 | ||
BU696808.1 | - | 5 | 512 | |||
X54511.1 | - | 6 | 652 |
Gene Symbol: 5730416F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017567.1 | - | 1 | 445 |