Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007603.3 | NP_031629.3 | 3 | 471 | ||
GenBank mRNA | AK145116.1 | - | 3 | 486 | ||
BC052167.1 | - | 3 | 372 | 6517565 | ||
CN700442.1 | - | 3 | 471 | |||
Y12582.1 | - | 3 | 380 |