Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007607.2 | NP_031633.1 | 5 | 588 | ||
GenBank mRNA | AK154754.1 | - | 5 | 554 | ||
AK165399.1 | - | 5 | 516 | |||
BC012704.1 | - | 5 | 513 | 4217743 |