Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007630.2 | NP_031656.2 | 4 | 425 | ||
GenBank mRNA | AK013312.1 | - | 4 | 394 | ||
AK076122.1 | - | 4 | 419 | |||
BC008247.1 | - | 4 | 341 | 3156950 | ||
CT010286.1 | - | 4 | 311 | |||
X66032.1 | - | 4 | 412 |