Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081635.1 | NP_001075104.1 | 7 | 1720 | ||
NM_001081636.1 | NP_001075105.1 | 7 | 1765 | |||
NM_007632.2 | NP_031658.1 | 5 | 1626 | |||
GenBank mRNA | AK083384.1 | - | 7 | 1706 | ||
AK159503.1 | - | 5 | 1552 | |||
AK161758.1 | - | 7 | 1644 | |||
AK168290.1 | - | 5 | 1554 | |||
AK188348.1 | - | 1 | 243 | |||
AK207692.1 | - | 1 | 291 | |||
BC004076.1 | - | 5 | 1518 | 3591985 | ||
BC005605.1 | - | 5 | 1517 | 3483584 |