Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 4 | 607 | ||
NM_001159556.1 | NP_001153028.1 | 4 | 467 | |||
NM_001159557.1 | NP_001153029.1 | 5 | 518 | |||
NM_001159558.1 | NP_001153030.1 | 5 | 634 | |||
NM_007643.4 | NP_031669.3 | 4 | 610 | |||
GenBank mRNA | AK052825.1 | - | 4 | 411 | ||
AK128908.1 | - | 5 | 411 | |||
AK150648.1 | - | 4 | 431 | |||
AK151355.1 | - | 4 | 446 | |||
AK152904.1 | - | 2 | 117 | |||
AK172162.1 | - | 5 | 456 | |||
BC010262.1 | - | 4 | 419 | 3481681 | ||
L23108.1 | - | 4 | 428 |