Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 17 | 3214 | ||
NM_018427.4 | NP_060897.3 | 18 | 3304 | |||
XM_005255376.1 | XP_005255433.1 | 16 | 3079 | |||
XM_005255377.4 | XP_005255434.1 | 15 | 3028 | |||
XM_011522540.2 | XP_011520842.1 | 13 | 2716 | |||
XM_011522541.1 | XP_011520843.1 | 17 | 3143 | |||
GenBank mRNA | AJ272050.1 | - | 18 | 3225 | ||
AL110238.1 | - | 1 | 458 | |||
BC036182.1 | - | 18 | 3219 | |||
BC071868.1 | - | 10 | 2539 |