Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001026214.2 | NP_001021385.1 | 6, 7 | 715 | ||
NM_001286049.1 | NP_001272978.1 | 5, 6 | 690 | |||
NM_001286058.1 | NP_001272987.1 | 4, 5 | 617 | |||
NM_007647.3 | NP_031673.2 | 5, 6 | 642 | |||
GenBank mRNA | AF006482.1 | - | 5, 6 | 650 | ||
AF136571.1 | - | 5, 6 | 676 | |||
AJ238636.1 | - | 5, 6 | 678 | |||
AK002618.1 | - | 5, 6 | 615 | |||
AK031581.1 | - | 6, 7 | 695 | |||
AK036641.1 | - | 6, 7 | 692 | |||
AK037736.1 | - | 5, 6 | 611 | |||
AK045828.1 | - | 5, 6 | 628 | |||
AK079267.1 | - | 4, 5 | 539 | |||
AK080265.1 | - | 6, 7 | 702 | |||
AK081435.1 | - | 6, 7 | 680 | |||
AK088455.1 | - | 5, 6 | 671 | |||
AK160950.1 | - | 3, 4 | 536 | |||
AK163688.1 | - | 5, 6 | 616 | |||
BC015247.1 | - | 4, 5 | 600 | 4242560 |