Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018441.5 | NP_060911.2 | 2, 3 | 341 | ||
XM_011511478.2 | XP_011509780.1 | 1, 2 | 142 | |||
GenBank mRNA | AF212234.1 | - | 2, 3 | 326 | ||
AF232009.1 | - | 2, 3 | 331 | |||
AJ250303.1 | - | 2, 3 | 327 | |||
AK296208.1 | - | 2, 3 | 315 | |||
AK315795.1 | - | 2, 3 | 300 | |||
BC002529.2 | - | 2, 3 | 305 | 3139219 | ||
BU151224.1 | - | 2, 3 | 341 | |||
BX471043.1 | - | 2, 3 | 335 |