Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007672.2 | NP_031698.2 | 2, 3 | 448 | ||
GenBank mRNA | AK148057.1 | - | 2, 3 | 432 | ||
AK171706.1 | - | 2, 3 | 403 | |||
BC117530.1 | - | 2, 3 | 337 | 40106297 | ||
BC118511.1 | - | 2, 3 | 337 | 40106298 | ||
BY713743.1 | - | 2, 3 | 448 | |||
U88588.1 | - | 3, 4 | 341 |