Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291310.1 | NP_001278239.1 | 2897 | |||
NM_018461.4 | NP_060931.2 | 1860 | ||||
NR_033191.2 | 1846 | |||||
XM_006717915.2 | XP_006717978.1 | 1276 | ||||
XM_017016406.1 | XP_016871895.1 | 1922 | ||||
XM_017016408.1 | XP_016871897.1 | 1128 | ||||
GenBank mRNA | AB040974.1 | - | 2335 | |||
AF220046.1 | - | 5 | 793 | |||
AK000402.1 | - | 6 | 1054 | |||
AK001483.1 | - | 2 | 496 | |||
BC047379.1 | - | 9 | 1860 | 5267510 | ||
BC058076.1 | - | 4 | 854 | |||
BC072402.1 | - | 6 | 1176 | 6048730 |