Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF012709.1 | - | 5 | 669 | ||
AK010718.1 | - | 5 | 631 | |||
AK011399.1 | - | 5 | 614 | |||
AK040313.1 | - | 6 | 796 | |||
AK041138.1 | - | 6 | 789 | |||
AK053763.1 | - | 5 | 638 | |||
AK089033.1 | - | 5 | 642 | |||
AK184358.1 | - | 2 | 150 | |||
BC011038.1 | - | 5 | 649 | 4018429 | ||
BC012280.1 | - | 7 | 742 |