Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007687.5 | NP_031713.1 | 2 | 312 | ||
GenBank mRNA | AK013943.1 | - | 2 | 241 | ||
AK020471.1 | - | 2 | 283 | |||
AK050651.1 | - | 243 | ||||
AK088633.1 | - | 2 | 248 | |||
AK088793.1 | - | 2 | 247 | |||
AK167795.1 | - | 2 | 243 | |||
BC046225.1 | - | 2 | 207 | 5691650 | ||
BC058726.1 | - | 2 | 240 | 5715707 | ||
BC094357.1 | - | 2 | 222 | 6820355 | ||
CX218255.1 | - | 2 | 348 | |||
D00472.1 | - | 2 | 261 |