Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321120.1 | NP_001308049.1 | 9 | 2517 | ||
NM_018488.3 | NP_060958.2 | 8 | 2354 | |||
XM_011525490.2 | XP_011523792.1 | 9 | 2555 | |||
XM_011525491.2 | XP_011523793.1 | 9 | 2552 | |||
GenBank mRNA | AF188703.1 | - | 8 | 2396 | ||
BC142620.1 | - | 9 | 2519 | 40147430 |