Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007691.5 | NP_031717.2 | 3 | 350 | ||
GenBank mRNA | AF016583.1 | - | 2 | 259 | ||
AK011258.1 | - | 3 | 294 | |||
AK033179.1 | - | 3 | 320 | |||
AK045336.1 | - | 3 | 292 | |||
AK166675.1 | - | 3 | 351 | |||
BC053394.1 | - | 3 | 286 | |||
BC064445.1 | - | 2 | 750 | |||
BC100386.1 | - | 2 | 425 | 30481390 |