Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007697.2 | NP_031723.2 | 4, 5 | 610 | ||
GenBank mRNA | AK040765.1 | - | 540 | |||
AK053039.1 | - | 3, 4 | 412 | |||
BC060216.1 | - | 4, 5 | 577 | |||
BC131670.1 | - | 4, 5 | 418 | 40110867 | ||
BC131671.1 | - | 4, 5 | 414 | 40110873 | ||
X94310.1 | - | 4, 5 | 484 |