Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301067.1 | NP_001287996.1 | 10 | 1351 | ||
NM_007598.4 | NP_031624.2 | 10 | 1335 | |||
GenBank mRNA | AK053351.1 | - | 10 | 1159 | ||
AK128889.1 | - | 10 | 1157 | |||
AK137222.1 | - | 10 | 1140 | |||
AK144094.1 | - | 10 | 1143 | |||
AK154002.1 | - | 10 | 1355 | |||
AK156556.1 | - | 10 | 1169 | |||
AK158150.1 | - | 9 | 1999 | |||
AK165565.1 | - | 10 | 1138 | |||
AK169652.1 | - | 10 | 1130 | |||
AK170901.1 | - | 10 | 1146 | |||
AK171388.1 | - | 10 | 1153 | |||
BC005446.1 | - | 9 | 1134 | 3600514 | ||
BC005472.1 | - | 9 | 1127 | 3590890 | ||
L12367.1 | - | 10 | 1161 |