Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042534.3 | NP_001035999.1 | 6 | 874 | ||
NM_001271395.1 | NP_001258324.1 | 6 | 867 | |||
NM_001271415.1 | NP_001258344.1 | 7 | 864 | |||
NM_007599.4 | NP_031625.2 | 6 | 721 | |||
GenBank mRNA | AK146622.1 | - | 6 | 725 | ||
AK149857.1 | - | 6 | 671 | |||
AK152818.1 | - | 6 | 699 | |||
AK159577.1 | - | 6 | 674 | |||
AK159736.1 | - | 6 | 676 | |||
AK165153.1 | - | 6 | 688 | |||
AK165586.1 | - | 4 | 517 | |||
BC003480.1 | - | 6 | 675 | 2811050 | ||
BC023101.1 | - | 6 | 663 | 3983574 | ||
X54511.1 | - | 7 | 863 |