Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007608.2 | NP_031634.2 | 3 | 483 | ||
GenBank mRNA | AI098626.1 | - | 3 | 399 | ||
AI314492.1 | - | 3 | 465 | |||
BC030174.1 | - | 3 | 461 | 5051543 | ||
X51971.1 | - | 3 | 475 |