Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007625.2 | NP_031651.2 | 5 | 1599 | ||
GenBank mRNA | AK040138.1 | - | 1 | 79 | ||
AK155152.1 | - | 1 | 703 | |||
AK155512.1 | - | 1 | 697 | |||
BC117801.1 | - | 5 | 1473 | 40092282 | ||
BC117802.1 | - | 5 | 1504 | 40092283 | ||
U63387.1 | - | 5 | 1511 |