Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cckbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028325.1 | - | 5 | 1743 | ||
AK053307.1 | - | 5 | 1603 | |||
BC103531.1 | - | 5 | 1713 | 40039474 | ||
BC103537.1 | - | 6 | 1662 | |||
BC103538.1 | - | 5 | 1713 | 40039475 |