Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007628.3 | NP_031654.2 | 3 | 465 | ||
GenBank mRNA | AK077114.1 | - | 3 | 471 | ||
BC120518.1 | - | 2 | 380 | 40129441 | ||
BC125436.1 | - | 2 | 380 | 40129951 | ||
X84311.1 | - | 3 | 464 |