Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007631.2 | NP_031657.1 | 5 | 2395 | ||
GenBank mRNA | AK053348.1 | - | 5 | 2303 | ||
AK164997.1 | - | 5 | 2340 | |||
AK169905.1 | - | 1 | 1232 | |||
AK170833.1 | - | 5 | 2279 | |||
S78355.1 | - | 5 | 2302 |