Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007637.2 | NP_031663.1 | 6 | 953 | ||
GenBank mRNA | AK077753.1 | - | 6 | 911 | ||
AK088184.1 | - | 6 | 887 | |||
AK129056.1 | - | 6 | 859 | |||
AK133857.1 | - | 6 | 911 | |||
AK136493.1 | - | 6 | 905 | |||
AK145445.1 | - | 6 | 922 | |||
AK146405.1 | - | 6 | 925 | |||
AK146425.1 | - | 6 | 905 | |||
AK150390.1 | - | 6 | 872 | |||
AK153911.1 | - | 6 | 929 | |||
AK167895.1 | - | 6 | 894 | |||
BC092233.1 | - | 6 | 923 | |||
BC094427.1 | - | 6 | 900 | 30537966 | ||
Z31555.1 | - | 6 | 953 |