Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 5 | 768 | ||
NM_001159556.1 | NP_001153028.1 | 5 | 628 | |||
NM_001159557.1 | NP_001153029.1 | 6 | 679 | |||
NM_001159558.1 | NP_001153030.1 | 6 | 795 | |||
NM_007643.4 | NP_031669.3 | 5 | 771 | |||
GenBank mRNA | AK052825.1 | - | 5 | 572 | ||
AK128908.1 | - | 6 | 572 | |||
AK150648.1 | - | 5 | 592 | |||
AK151355.1 | - | 5 | 607 | |||
AK152904.1 | - | 3 | 278 | |||
AK172162.1 | - | 6 | 617 | |||
BC010262.1 | - | 5 | 580 | 3481681 | ||
L23108.1 | - | 5 | 589 |