Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001026214.2 | NP_001021385.1 | 7 | 721 | ||
NM_001286049.1 | NP_001272978.1 | 6 | 696 | |||
NM_001286058.1 | NP_001272987.1 | 5 | 623 | |||
NM_007647.3 | NP_031673.2 | 6 | 648 | |||
GenBank mRNA | AF006482.1 | - | 6 | 656 | ||
AF136571.1 | - | 6 | 682 | |||
AJ238636.1 | - | 6 | 684 | |||
AK002618.1 | - | 6 | 621 | |||
AK031581.1 | - | 7 | 701 | |||
AK036641.1 | - | 7 | 698 | |||
AK037736.1 | - | 6 | 617 | |||
AK045828.1 | - | 6 | 634 | |||
AK079267.1 | - | 5 | 545 | |||
AK080265.1 | - | 7 | 708 | |||
AK081435.1 | - | 7 | 686 | |||
AK088455.1 | - | 6 | 677 | |||
AK160950.1 | - | 4 | 542 | |||
AK163688.1 | - | 6 | 622 | |||
BC015247.1 | - | 5 | 606 | 4242560 |