Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007651.3 | NP_031677.1 | 8 | 1687 | ||
GenBank mRNA | AK088232.1 | - | 1 | 823 | ||
AK156510.1 | - | 8 | 1595 | |||
AK160326.1 | - | 8 | 1585 | |||
BC021310.1 | - | 8 | 1600 | 5010267 | ||
BC052905.1 | - | 8 | 1548 | 30078163 |