Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042580.1 | NP_001036045.1 | 8 | 912 | ||
NM_001282966.1 | NP_001269895.1 | 8 | 956 | |||
NM_007653.3 | NP_031679.1 | 8 | 871 | |||
GenBank mRNA | AF159427.1 | - | 8 | 792 | ||
AK028220.1 | - | 8 | 809 | |||
AK150647.1 | - | 8 | 866 | |||
AK151702.1 | - | 8 | 837 | |||
AK151963.1 | - | 8 | 811 | |||
AK152072.1 | - | 8 | 833 | |||
AK152372.1 | - | 8 | 837 | |||
AK159875.1 | - | 8 | 834 | |||
AK163785.1 | - | 8 | 833 | |||
AK166373.1 | - | 1 | 2881 | |||
AK166543.1 | - | 8 | 832 | |||
AK166766.1 | - | 8 | 831 | |||
AK170344.1 | - | 8 | 866 | |||
AK184072.1 | - | 2 | 142 | |||
AK184717.1 | - | 2 | 142 | |||
AK211715.1 | - | 3 | 294 | |||
BC008108.1 | - | 8 | 818 | 3157993 | ||
BC012212.1 | - | 8 | 818 | 3157993 | ||
BC083161.1 | - | 8 | 803 | 6530532 | ||
CJ210573.1 | - | 3 | 349 | |||
D16432.1 | - | 8 | 821 |