Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007659.3 | NP_031685.2 | 8 | 2383 | ||
GenBank mRNA | AK030231.1 | - | 8 | 2384 |