Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF012709.1 | - | 5 | 745 | ||
AK010718.1 | - | 5 | 707 | |||
AK011399.1 | - | 5 | 690 | |||
AK040313.1 | - | 6 | 872 | |||
AK041138.1 | - | 6 | 865 | |||
AK053763.1 | - | 5 | 714 | |||
AK089033.1 | - | 5 | 718 | |||
BC011038.1 | - | 5 | 725 | 4018429 | ||
BC012280.1 | - | 7 | 818 |