Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007683.3 | NP_031709.2 | 6, 7 | 767 | ||
GenBank mRNA | AK137803.1 | - | 6, 7 | 736 | ||
AK143566.1 | - | 6, 7 | 768 | |||
BC150700.1 | - | 6, 7 | 563 | 9087611 | ||
U03113.1 | - | 6, 7 | 653 |