Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007691.5 | NP_031717.2 | 3 | 389 | ||
GenBank mRNA | AF016583.1 | - | 2 | 298 | ||
AK011258.1 | - | 3 | 333 | |||
AK033179.1 | - | 3 | 359 | |||
AK045336.1 | - | 3 | 331 | |||
AK166675.1 | - | 3 | 390 | |||
BC053394.1 | - | 3 | 325 | |||
BC064445.1 | - | 2 | 789 | |||
BC100386.1 | - | 2 | 464 | 30481390 |