Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007692.6 | NP_031718.1 | 6, 7 | 1071 | ||
GenBank mRNA | AB011000.1 | - | 6, 7 | 1043 | ||
AK078101.1 | - | 6, 7 | 1065 | |||
AK170077.1 | - | 5, 6 | 752 | |||
BC068195.1 | - | 6, 7 | 589 | 4164116 | ||
BC069925.1 | - | 4, 5 | 1392 |