Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162410.1 | NP_001155882.1 | 6 | 616 | ||
NM_007700.2 | NP_031726.2 | 6 | 616 | |||
GenBank mRNA | AK018671.1 | - | 6 | 595 | ||
AK035326.1 | - | 6 | 573 | |||
AK150039.1 | - | 6 | 595 | |||
AK155328.1 | - | 6 | 596 | |||
AK155393.1 | - | 6 | 617 | |||
AK164236.1 | - | 6 | 617 | |||
BC018243.1 | - | 6 | 608 | 4511275 | ||
U12473.1 | - | 6 | 584 |