Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111110.2 | NP_001104580.1 | 4 | 607 | ||
NM_001284519.1 | NP_001271448.1 | 5 | 769 | |||
NM_001284520.1 | NP_001271449.1 | 3 | 262 | |||
NM_007717.5 | NP_031743.3 | 3 | 262 | |||
GenBank mRNA | AB061276.1 | - | 4 | 591 | ||
AB061277.1 | - | 5 | 753 | |||
AK040274.1 | - | 2 | 212 | |||
AK083394.1 | - | 2 | 296 | |||
AK145110.1 | - | 2 | 410 | |||
AK145199.1 | - | 3 | 263 | |||
AK166486.1 | - | 3 | 263 | |||
BC028474.1 | - | 4 | 617 | |||
BC055079.1 | - | 2 | 259 | 30077348 | ||
CA577904.1 | - | 2 | 247 | |||
D21826.1 | - | 2 | 234 |