Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007725.2 | NP_031751.1 | 2 | 176 | ||
GenBank mRNA | AK051845.1 | - | 2 | 177 | ||
AK189304.1 | - | 2 | 104 | |||
AK218463.1 | - | 1 | 38 | |||
BC008181.1 | - | 2 | 94 | 3589470 | ||
BC009144.1 | - | 2 | 131 | 3594856 | ||
BC018482.1 | - | 2 | 146 | 3589158 |