Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019069.3 | NP_061942.2 | 1 | 1863 | ||
GenBank mRNA | BC025762.1 | - | 1 | 1862 | ||
BC043494.1 | - | 1 | 1452 | 6056052 |