Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037859.2 | NP_001032948.2 | 4 | 695 | ||
GenBank mRNA | AK004947.1 | - | 3 | 505 | ||
AK041299.1 | - | 3 | 483 | |||
AK076215.1 | - | 3 | 502 | |||
AK079247.1 | - | 3 | 512 | |||
AK080326.1 | - | 3 | 491 | |||
AK089498.1 | - | 3 | 485 | |||
AK143545.1 | - | 3 | 496 | |||
AK146069.1 | - | 4 | 696 | |||
AK154128.1 | - | 3 | 510 | |||
AK154556.1 | - | 3 | 506 | |||
AK154653.1 | - | 3 | 506 | |||
AK154699.1 | - | 3 | 506 | |||
AK155488.1 | - | 3 | 515 | |||
AK155565.1 | - | 3 | 525 | |||
AK155576.1 | - | 512 | ||||
AK155640.1 | - | 3 | 510 | |||
AK155654.1 | - | 3 | 510 | |||
AK155858.1 | - | 3 | 513 | |||
AK156550.1 | - | 3 | 530 | |||
AK171241.1 | - | 3 | 545 | |||
AK171386.1 | - | 3 | 510 | |||
AK171543.1 | - | 3 | 485 | |||
BC036343.1 | - | 3 | 482 | 4948135 | ||
BC043054.1 | - | 3 | 486 | 6418718 | ||
BC050024.1 | - | 3 | 552 | |||
BC066863.1 | - | 4 | 604 | 30436119 | ||
X06368.1 | - | 3 | 488 | |||
X68932.1 | - | 3 | 488 |