Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007780.4 | NP_031806.3 | 9, 10 | 1430 | ||
GenBank mRNA | AF483496.1 | - | 8, 9 | 1172 | ||
AF483497.1 | - | 8, 9 | 1172 | |||
AK133420.1 | - | 1374 | ||||
AK152608.1 | - | 9, 10 | 1370 | |||
AK154286.1 | - | 9, 10 | 1367 | |||
AK155109.1 | - | 9, 10 | 1401 | |||
AK155215.1 | - | 9, 10 | 1310 | |||
AK155699.1 | - | 9, 10 | 1367 | |||
AK155803.1 | - | 9, 10 | 1367 | |||
AK155822.1 | - | 9, 10 | 1370 | |||
AK155885.1 | - | 9, 10 | 1371 | |||
AK171477.1 | - | 9, 10 | 1367 | |||
BC113202.1 | - | 10, 11 | 1441 | 40048156 | ||
M34397.1 | - | 10, 11 | 1459 |