Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007798.3 | NP_031824.1 | 4 | 365 | ||
GenBank mRNA | AK002456.1 | - | 4 | 314 | ||
AK083393.1 | - | 4 | 365 | |||
AK147192.1 | - | 4 | 312 | |||
AK149884.1 | - | 4 | 314 | |||
AK151790.1 | - | 4 | 312 | |||
AK153047.1 | - | 4 | 311 | |||
AK159992.1 | - | 4 | 350 | |||
AK167361.1 | - | 4 | 346 | |||
AK170959.1 | - | 4 | 338 | |||
AK190209.1 | - | 3 | 163 | |||
AK190539.1 | - | 3 | 163 | |||
AK191055.1 | - | 3 | 163 | |||
AK196955.1 | - | 3 | 163 | |||
AK198213.1 | - | 3 | 163 | |||
AK199268.1 | - | 3 | 164 | |||
AK201893.1 | - | 3 | 163 | |||
AK202291.1 | - | 3 | 163 | |||
AK203639.1 | - | 3 | 163 | |||
BC006656.1 | - | 4 | 356 | 3500700 | ||
M14222.1 | - | 3 | 270 | |||
S69034.1 | - | 4 | 361 |