Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301284.1 | NP_001288213.1 | 5 | 540 | ||
NM_007806.3 | NP_031832.2 | 6 | 624 | |||
GenBank mRNA | AK018713.1 | - | 5 | 509 | ||
AK021200.1 | - | 6 | 612 | |||
AK078854.1 | - | 6 | 580 | |||
AK131580.1 | - | 6 | 595 | |||
AK152414.1 | - | 6 | 615 | |||
AK171748.1 | - | 6 | 593 | |||
AK190653.1 | - | 1 | 72 | |||
BC026791.1 | - | 6 | 575 | 3980538 | ||
BC060635.1 | - | 5 | 4626 | |||
CF738398.1 | - | 6 | 607 | |||
EU791539.1 | - | 6 | 554 | |||
M31775.1 | - | 6 | 547 |