Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007807.5 | NP_031833.3 | 2 | 175 | ||
GenBank mRNA | AK033570.1 | - | 2 | 119 | ||
AK053920.1 | - | 2 | 119 | |||
AK079926.1 | - | 2 | 122 | |||
AK152257.1 | - | 2 | 146 | |||
AK153154.1 | - | 2 | 146 | |||
AK153259.1 | - | 2 | 146 | |||
AK154126.1 | - | 2 | 175 | |||
AK171526.1 | - | 2 | 161 | |||
BC042838.1 | - | 2 | 142 | |||
BC071229.1 | - | 2 | 130 | 4979804 | ||
BY194282.1 | - | 2 | 175 | |||
FJ168469.1 | - | 2 | 164 | |||
U43384.1 | - | 2 | 101 |